Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024734.3 | NP_079010.2 | 4 | 402 | ||
XM_011537158.1 | XP_011535460.1 | 4 | 398 | |||
XM_011537159.2 | XP_011535461.1 | 4 | 398 | |||
XM_017021646.1 | XP_016877135.1 | 4 | 568 | |||
XM_017021647.1 | XP_016877136.1 | 4 | 398 | |||
GenBank mRNA | AB047979.1 | - | 4 | 359 | ||
AK025226.1 | - | 4 | 345 | |||
AK314315.1 | - | 4 | 400 | |||
AL117526.1 | - | 4 | 254 | |||
BC068482.1 | - | 4 | 351 | 4375469 | ||
CB155696.1 | - | 4 | 402 |