Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024762.3 | NP_079038.2 | 3 | 1269 | ||
XM_005259267.3 | XP_005259324.1 | 3 | 1447 | |||
GenBank mRNA | AK025256.1 | - | 1 | 573 | ||
AK097041.1 | - | 3 | 1269 | |||
AK225219.1 | - | 1 | 573 | |||
AK293445.1 | - | 3 | 1448 | |||
BC017460.1 | - | 1 | 707 | |||
BC046237.1 | - | 1 | 707 | |||
BC062982.1 | - | 1 | 707 | |||
BC082767.1 | - | 1 | 707 |
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 2715 | ||
XM_017027197.1 | XP_016882686.1 | 2 | 2360 | |||
GenBank mRNA | AK300483.1 | - | 3 | 2681 |