Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076786.2 | NP_001070254.1 | 10 | 5101 | ||
XM_017018327.1 | XP_016873816.1 | 10 | 5453 | |||
XM_017018328.1 | XP_016873817.1 | 10 | 5453 | |||
XM_017018329.1 | XP_016873818.1 | 8 | 5906 | |||
XM_017018330.1 | XP_016873819.1 | 9 | 4851 | |||
GenBank mRNA | AK025577.1 | - | 7 | 1828 | ||
AK125902.1 | - | 7 | 2649 | |||
AK126023.1 | - | 11 | 4405 | |||
AL834141.1 | - | 7 | 3797 | |||
BC112935.1 | - | 10 | 5101 |