Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105248.1 | NP_001098718.1 | 10 | 2472 | ||
NM_001105249.1 | NP_001098719.1 | 10 | 2472 | |||
NM_001261841.1 | NP_001248770.1 | 10 | 2427 | |||
NM_001308161.1 | NP_001295090.1 | 10 | 2265 | |||
NM_024780.4 | NP_079056.2 | 6 | 1242 | |||
GenBank mRNA | AK093944.1 | - | 3 | 299 | ||
AK097718.1 | - | 6 | 1207 | |||
AK131202.1 | - | 7 | 962 | |||
AK299055.1 | - | 10 | 2265 | |||
AK304431.1 | - | 10 | 2427 | |||
AY236494.1 | - | 6 | 1242 | |||
AY263164.1 | - | 6 | 985 | |||
AY358155.1 | - | 8 | 1821 | |||
BC027602.1 | - | 6 | 1197 | 4839111 | ||
BC038118.2 | - | 6 | 937 | 5265527 | ||
CR749359.1 | - | 8 | 1127 |