Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFAP69 SIRNA Assays ›
Gene Symbol:
CFAP69
Gene Name:
cilia and flagella associated protein 69
Gene Aliases:
C7orf63, FAP69
Chromosome Location:
Chr.7: 90245174 - 90320964 on Build GRCh38
UniGene ID:
Hs.521014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36474
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFAP69
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039706.2
NP_001034795.2
17
2158
NM_001160138.1
NP_001153610.1
17
2104
XM_011516577.2
XP_011514879.1
17
2137
XM_011516578.2
XP_011514880.1
17
2137
XM_017012624.1
XP_016868113.1
17
2083
XM_017012625.1
XP_016868114.1
17
2137
XM_017012626.1
XP_016868115.1
17
2137
XM_017012627.1
XP_016868116.1
17
2137
XM_017012628.1
XP_016868117.1
17
2137
XM_017012629.1
XP_016868118.1
16
2027
XM_017012630.1
XP_016868119.1
17
2137
XM_017012631.1
XP_016868120.1
15
1685
XM_017012632.1
XP_016868121.1
17
2137
XM_017012633.1
XP_016868122.1
15
4714
XM_017012634.1
XP_016868123.1
15
1852
XM_017012635.1
XP_016868124.1
13
1548
XM_017012636.1
XP_016868125.1
12
1489
XM_017012637.1
XP_016868126.1
13
4654
XM_017012638.1
XP_016868127.1
13
1495
XM_017012639.1
XP_016868128.1
13
1530
XM_017012640.1
XP_016868129.1
10
1134
GenBank mRNA
AK024715.1
-
6
656
AK093991.1
-
15
1940
AK302636.1
-
17
2063
AK303034.1
-
17
2137
AL833446.1
-
11
1351
BC036351.1
-
18
2322
BC132771.1
-
11
1380
BC140742.1
-
17
2044
9021610
BC141834.1
-
17
2104
40148850
BC142655.1
-
17
2104
40148060
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762734, XM_006236074, XM_003749688, XM_008762732, XM_008762733.This siRNA also targets the following transcripts in Mouse: NM_172447, AK164190, AK039313, BC062906, AK076835, and others.
Panther Classification:
Molecular Function -
structural protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0