Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143850.2 | NP_001137322.1 | 14, 15 | 1731 | ||
NM_024809.4 | NP_079085.2 | 14, 15 | 1734 | |||
XM_005253623.2 | XP_005253680.1 | 13, 14 | 1599 | |||
XM_006719605.3 | XP_006719668.1 | 14, 15 | 1734 | |||
XM_017019974.1 | XP_016875463.1 | 13, 14 | 1596 | |||
XM_017019975.1 | XP_016875464.1 | 13, 14 | 1609 | |||
GenBank mRNA | AK023037.1 | - | 3, 4 | 290 | ||
AK056924.1 | - | 14, 15 | 1732 | |||
AK292153.1 | - | 14, 15 | 1731 | |||
BC009112.1 | - | 14, 15 | 1703 | 3463475 | ||
DQ278870.1 | - | 14, 15 | 1732 |