Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001897.4 | NP_001888.2 | 4, 5 | 4366 | ||
GenBank mRNA | AY359468.1 | - | 4, 5 | 4273 | ||
X96753.1 | - | 4, 5 | 4273 |