Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 2 | 371 | ||
NM_024818.4 | NP_079094.1 | 2 | 1221 | |||
NM_198329.3 | NP_938143.1 | 2 | 244 | |||
GenBank mRNA | AB154406.1 | - | 2 | 180 | ||
AK026904.1 | - | 2 | 414 | |||
AK027563.1 | - | 1 | 13 | |||
AK056238.1 | - | 2 | 1222 | |||
AK129616.1 | - | 3 | 280 | |||
AL136757.1 | - | 2 | 234 | |||
AY253672.1 | - | 2 | 373 | |||
BC009737.1 | - | 2 | 350 | 3879061 | ||
BC048015.1 | - | 2 | 328 | |||
BM541647.1 | - | 2 | 707 | |||
DA785394.1 | - | 2 | 334 | |||
DB082596.1 | - | 2 | 423 | |||
DB457433.1 | - | 2 | 245 |