Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024821.3 | NP_079097.1 | 6, 7 | 721 | ||
XM_005261748.3 | XP_005261805.1 | 7, 8 | 811 | |||
GenBank mRNA | AK026002.1 | - | 6, 7 | 664 | ||
BC017693.2 | - | 6, 7 | 646 | 4403280 | ||
CR456484.1 | - | 6, 7 | 646 |