Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024821.3 | NP_079097.1 | 6 | 696 | ||
XM_005261748.3 | XP_005261805.1 | 7 | 786 | |||
GenBank mRNA | AK026002.1 | - | 6 | 639 | ||
BC017693.2 | - | 6 | 621 | 4403280 | ||
CR456484.1 | - | 6 | 621 |