Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36557
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
15
2307
NM_001160104.1
NP_001153576.1
15
2292
NM_001326295.1
NP_001313224.1
13
1917
NM_001326296.1
NP_001313225.1
14
2232
NM_001326297.1
NP_001313226.1
15
2491
NM_001326298.1
NP_001313227.1
12
1842
NM_001326299.1
NP_001313228.1
14
2217
NM_001326300.1
NP_001313229.1
14
2195
NM_001326301.1
NP_001313230.1
14
2416
NM_001326302.1
NP_001313231.1
14
2192
NM_001326303.1
NP_001313232.1
12
2020
NM_001326304.1
NP_001313233.1
12
1799
NM_001326305.1
NP_001313234.1
13
2120
NM_001326306.1
NP_001313235.1
11
1683
NM_001326307.1
NP_001313236.1
14
2235
NM_001326308.1
NP_001313237.1
12
1802
NM_001326309.1
NP_001313238.1
11
1727
NM_001326310.1
NP_001313239.1
15
2295
NM_001326311.1
NP_001313240.1
11
1724
NM_001326312.1
NP_001313241.1
14
2220
NM_001326313.1
NP_001313242.1
11
1686
NM_001326314.1
NP_001313243.1
13
2117
NM_001326315.1
NP_001313244.1
15
2488
NM_001326316.1
NP_001313245.1
15
2155
NM_024824.4
NP_079100.2
15
2310
NM_207660.3
NP_997543.1
12
1839
NM_207661.2
NP_997544.1
13
1765
NM_207662.3
NP_997545.2
5
1136
NR_136936.1
14
2150
XM_005268067.4
XP_005268124.1
15
2290
XM_005268068.4
XP_005268125.1
14
2230
XM_005268069.4
XP_005268126.1
13
1912
XM_005268070.4
XP_005268127.1
13
1909
XM_005268071.4
XP_005268128.1
12
1822
XM_005268073.3
XP_005268130.1
7
1532
XM_006720257.2
XP_006720320.1
6
1457
XM_011537160.2
XP_011535462.1
15
2418
XM_011537161.2
XP_011535463.1
14
2149
XM_017021648.1
XP_016877137.1
14
2227
XM_017021649.1
XP_016877138.1
14
2215
XM_017021650.1
XP_016877139.1
14
2212
GenBank mRNA
AF155107.1
-
4
398
AF474376.1
-
4
1005
AK021868.1
-
12
1686
AK090809.1
-
1
1370
AK302136.1
-
12
1657
AK303123.1
-
14
1758
AK304275.1
-
15
2163
AL834215.1
-
13
1765
AY578060.1
-
15
2194
AY578061.1
-
12
1778
AY578062.1
-
13
1765
AY578063.1
-
7
843
BC011793.1
-
15
2176
4298961
BC023641.2
-
11
1813
BC027607.2
-
5
1122
BX248265.1
-
11
1829
BX248281.1
-
7
911
BX248769.1
-
2
200
BX640716.1
-
15
2189
BX640876.1
-
4
503
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0