Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039580.1 | NP_001034669.1 | 9, 10 | 1546 | ||
XM_006714306.3 | XP_006714369.1 | 9, 10 | 1691 | |||
XM_011532253.1 | XP_011530555.1 | 9, 10 | 1749 | |||
XM_011532254.2 | XP_011530556.1 | 9, 10 | 1658 | |||
XM_011532255.2 | XP_011530557.1 | 8, 9 | 1397 | |||
XM_017008616.1 | XP_016864105.1 | 9, 10 | 1746 | |||
XM_017008617.1 | XP_016864106.1 | 9, 10 | 1563 | |||
XM_017008618.1 | XP_016864107.1 | 9, 10 | 1524 | |||
GenBank mRNA | AK024730.1 | - | 6, 7 | 945 | ||
AK301075.1 | - | 9, 10 | 1338 | |||
AY690636.1 | - | 9, 10 | 1546 | |||
BC022852.1 | - | 9 | 1371 | |||
BC128540.1 | - | 9, 10 | 1458 | |||
BC128541.1 | - | 9, 10 | 1461 | |||
BC146864.1 | - | 9, 10 | 1458 | 9056792 |