Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136041.2 | NP_001129513.1 | 4 | 388 | ||
NM_024827.3 | NP_079103.2 | 4 | 489 | |||
XM_005265491.1 | XP_005265548.1 | 4 | 454 | |||
XM_006713338.1 | XP_006713401.1 | 4 | 489 | |||
XM_011534131.1 | XP_011532433.1 | 4 | 577 | |||
XM_011534135.1 | XP_011532437.1 | 3 | 296 | |||
XM_017007234.1 | XP_016862723.1 | 4 | 577 | |||
GenBank mRNA | AK025426.1 | - | 4 | 380 | ||
AK025890.1 | - | 4 | 380 | |||
AK293223.1 | - | 4 | 321 | |||
AL137362.1 | - | 1 | 20 | |||
AL834223.1 | - | 4 | 747 | |||
BC009676.1 | - | 3 | 365 | 3906049 | ||
BG708402.1 | - | 4 | 459 | |||
DB025431.1 | - | 4 | 489 |