Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 10, 11 | 1690 | ||
XM_005257689.2 | XP_005257746.1 | 10, 11 | 1672 | |||
XM_011525265.2 | XP_011523567.1 | 9, 10 | 1435 | |||
XM_011525266.1 | XP_011523568.1 | 9, 10 | 1558 | |||
XM_017025107.1 | XP_016880596.1 | 9, 10 | 1403 | |||
GenBank mRNA | AF126964.1 | - | 9, 10 | 1421 | ||
AJ404670.2 | - | 2, 3 | 247 | |||
AK026214.1 | - | 2, 3 | 324 | |||
AK124053.1 | - | 4, 5 | 2061 | |||
AK124084.1 | - | 4, 5 | 1606 | |||
AK290687.1 | - | 9, 10 | 1446 | |||
AK301614.1 | - | 7, 8 | 978 | |||
BC137491.1 | - | 10, 11 | 1556 | 9021491 | ||
CD557886.1 | - | 3, 4 | 520 |