Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 7 | 1051 | ||
NM_144722.3 | NP_653323.1 | 7 | 1051 | |||
XM_005248376.3 | XP_005248433.1 | 7 | 1156 | |||
XM_005248377.3 | XP_005248434.1 | 7 | 1156 | |||
XM_005248378.3 | XP_005248435.1 | 7 | 1145 | |||
XM_011514135.2 | XP_011512437.1 | 7 | 1161 | |||
XM_011514136.2 | XP_011512438.1 | 7 | 1160 | |||
XM_011514137.2 | XP_011512439.1 | 7 | 1160 | |||
XM_011514138.2 | XP_011512440.1 | 7 | 1161 | |||
XM_011514139.2 | XP_011512441.1 | 7 | 1160 | |||
XM_011514140.2 | XP_011512442.1 | 6 | 895 | |||
XM_011514141.2 | XP_011512443.1 | 8 | 1359 | |||
XM_011514142.1 | XP_011512444.1 | 7 | 1156 | |||
XM_017009880.1 | XP_016865369.1 | 6 | 992 | |||
XM_017009881.1 | XP_016865370.1 | 7 | 2015 | |||
XM_017009882.1 | XP_016865371.1 | 7 | 1149 | |||
GenBank mRNA | AK058124.1 | - | 7 | 976 | ||
BC110984.1 | - | 7 | 966 | |||
EF553528.1 | - | 7 | 1001 |