Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHDDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243565.1 | NP_001230494.1 | 5, 6 | 575 | ||
NM_001319959.1 | NP_001306888.1 | 6, 7 | 715 | |||
NM_024887.3 | NP_079163.2 | 6, 7 | 692 | |||
NM_205861.2 | NP_995583.1 | 6, 7 | 692 | |||
XM_006710912.2 | XP_006710975.1 | 6, 7 | 751 | |||
XM_006710913.2 | XP_006710976.1 | 6, 7 | 630 | |||
XM_006710914.2 | XP_006710977.1 | 6, 7 | 627 | |||
XM_006710916.2 | XP_006710979.1 | 6, 7 | 741 | |||
XM_006710917.2 | XP_006710980.1 | 6, 7 | 694 | |||
XM_006710918.2 | XP_006710981.1 | 6, 7 | 650 | |||
XM_011542183.2 | XP_011540485.1 | 6, 7 | 727 | |||
XM_011542184.2 | XP_011540486.1 | 6, 7 | 748 | |||
XM_017002379.1 | XP_016857868.1 | 6, 7 | 630 | |||
XM_017002380.1 | XP_016857869.1 | 6, 7 | 727 | |||
XM_017002381.1 | XP_016857870.1 | 6, 7 | 741 | |||
XM_017002382.1 | XP_016857871.1 | 6, 7 | 650 | |||
GenBank mRNA | AB090852.1 | - | 5, 6 | 553 | ||
AK023164.1 | - | 6, 7 | 622 | |||
AK308187.1 | - | 6, 7 | 634 | |||
BC003643.2 | - | 5, 6 | 597 | 3532466 | ||
BC004117.2 | - | 5, 6 | 517 | 3843062 | ||
BX648507.1 | - | 6, 7 | 717 | |||
GU727641.1 | - | 5, 6 | 597 |