Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001899.2 | NP_001890.1 | 3 | 410 | ||
GenBank mRNA | BC065714.1 | - | 3 | 427 | 30327736 | |
BC074952.2 | - | 3 | 383 | 30915278 | ||
BC074953.2 | - | 3 | 383 | 30915500 |
Gene Symbol: CST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001898.2 | NP_001889.2 | 3 | 425 | ||
GenBank mRNA | BC021225.2 | - | 3 | 423 | 4298603 | |
J03870.1 | - | 3 | 424 |