Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 6 | 1070 | ||
NM_001193334.1 | NP_001180263.1 | 5 | 803 | |||
NM_024911.6 | NP_079187.3 | 6 | 1131 | |||
XM_011542191.2 | XP_011540493.1 | 6 | 1054 | |||
XM_011542192.2 | XP_011540494.1 | 6 | 1064 | |||
XM_017002390.1 | XP_016857879.1 | 6 | 898 | |||
GenBank mRNA | AK026744.1 | - | 3 | 212 | ||
AK074984.1 | - | 6 | 1053 | |||
AK301613.1 | - | 5 | 663 | |||
AK309779.1 | - | 7 | 1010 | |||
AY359035.1 | - | 6 | 1113 | |||
BC007211.1 | - | 5 | 669 | |||
BC110826.1 | - | 6 | 1032 | 6183768 | ||
BC137109.1 | - | 6 | 928 | 9021106 | ||
BC137113.1 | - | 6 | 928 | 9021110 | ||
BX538320.1 | - | 6 | 1070 | |||
BX648748.1 | - | 5 | 801 | |||
DQ323735.1 | - | 6 | 1100 |