Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024949.5 | NP_079225.5 | 14 | 2374 | ||
XM_011532269.2 | XP_011530571.1 | 14 | 3007 | |||
GenBank mRNA | AJ566366.1 | - | 1 | 56 | ||
AK074260.1 | - | 1 | 86 | |||
AK126057.1 | - | 13 | 2039 | |||
AK127061.1 | - | 7 | 1286 | |||
BC053873.1 | - | 5 | 803 | |||
BC107683.1 | - | 5 | 971 | |||
BX647378.1 | - | 12 | 1965 | |||
BX647704.1 | - | 2 | 101 |