Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 10 | 1522 | ||
NM_001102367.2 | NP_001095837.1 | 9 | 1288 | |||
NM_001102368.2 | NP_001095838.1 | 8 | 1121 | |||
NM_001102369.2 | NP_001095839.1 | 10 | 1241 | |||
NM_001286470.1 | NP_001273399.1 | 11 | 1816 | |||
NM_001286471.1 | NP_001273400.1 | 8 | 1357 | |||
NM_001286472.1 | NP_001273401.1 | 10 | 1292 | |||
NM_024952.7 | NP_079228.4 | 8 | 1351 | |||
GenBank mRNA | AB210031.1 | - | 9 | 1022 | ||
AK022564.1 | - | 8 | 922 | |||
AK023200.1 | - | 10 | 1039 | |||
AK024603.1 | - | 2 | 147 | |||
AK091394.1 | - | 9 | 711 | |||
AK097294.1 | - | 8 | 1071 | |||
AK098752.1 | - | 8 | 1072 | |||
AK123532.1 | - | 11 | 1459 | |||
AK222531.1 | - | 2 | 147 | |||
AK308857.1 | - | 6 | 784 | |||
AY358516.1 | - | 8 | 1357 | |||
BC010614.2 | - | 9 | 1237 | 4157432 | ||
BC065558.1 | - | 10 | 1145 | 6138321 | ||
BX161393.1 | - | 8 | 1050 | |||
BX247989.1 | - | 10 | 1220 | |||
BX248294.1 | - | 6 | 723 | |||
BX248782.1 | - | 9 | 1071 | |||
BX537970.1 | - | 8 | 1068 | |||
CR457289.1 | - | 6 | 769 |