Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321213.1 | NP_001308142.1 | 2 | 341 | ||
NM_024963.5 | NP_079239.3 | 2 | 341 | |||
XM_011515531.2 | XP_011513833.1 | 2 | 356 | |||
XM_011515532.2 | XP_011513834.1 | 2 | 356 | |||
GenBank mRNA | AK057042.1 | - | 2 | 295 | ||
AK294949.1 | - | 2 | 335 | |||
BC006426.1 | - | 2 | 311 | 4026501 | ||
BC024256.1 | - | 2 | 311 | 3954975 | ||
DC307306.1 | - | 2 | 341 |