Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302627.1 | NP_001289556.1 | 4 | 505 | ||
NM_025031.2 | NP_079307.2 | 3 | 361 | |||
GenBank mRNA | AK024728.1 | - | 3 | 361 | ||
BC113679.1 | - | 3 | 352 | 8322731 | ||
BC113681.1 | - | 3 | 352 | 8322733 | ||
DA711092.1 | - | 4 | 468 |