Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286496.1 | NP_001273425.1 | 11 | 1776 | ||
NM_001286497.1 | NP_001273426.1 | 11 | 1752 | |||
NM_001286499.1 | NP_001273428.1 | 11 | 1776 | |||
NM_025049.3 | NP_079325.2 | 11 | 1752 | |||
XM_011522083.2 | XP_011520385.1 | 11 | 1868 | |||
XM_011522084.2 | XP_011520386.1 | 11 | 1700 | |||
GenBank mRNA | AB185926.1 | - | 11 | 1752 | ||
AB185927.1 | - | 11 | 1752 | |||
AF108138.1 | - | 7 | 856 | |||
AY498716.1 | - | 10 | 1676 | |||
BC033254.1 | - | 5 | 617 | |||
BC137503.1 | - | 11 | 1690 | 9021503 | ||
BC137504.1 | - | 11 | 1690 | 9021504 | ||
DQ437529.1 | - | 10 | 1657 | |||
EU084033.1 | - | 11 | 1747 |