Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARMC9 SIRNA Assays ›
Gene Symbol:
ARMC9
Gene Name:
armadillo repeat containing 9
Gene Aliases:
ARM, KU-MEL-1, NS21
Chromosome Location:
Chr.2: 231198546 - 231394991 on Build GRCh38
UniGene ID:
Hs.162411
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37063
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARMC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271466.2
NP_001258395.1
6
650
NM_001291656.1
NP_001278585.1
7
803
NM_025139.5
NP_079415.3
7
805
XM_011511905.1
XP_011510207.1
7
723
XM_011511906.1
XP_011510208.1
7
723
XM_011511907.1
XP_011510209.1
7
723
XM_011511908.2
XP_011510210.1
7
723
XM_011511909.2
XP_011510211.1
7
723
XM_011511910.1
XP_011510212.1
7
723
XM_011511911.1
XP_011510213.1
7
723
XM_011511912.1
XP_011510214.1
7
723
XM_011511913.2
XP_011510215.1
7
723
XM_011511914.2
XP_011510216.1
7
723
XM_011511915.2
XP_011510217.1
7
723
XM_011511916.2
XP_011510218.1
7
723
XM_017005018.1
XP_016860507.1
7
981
XM_017005019.1
XP_016860508.1
7
899
XM_017005020.1
XP_016860509.1
7
981
XM_017005021.1
XP_016860510.1
7
723
XM_017005022.1
XP_016860511.1
7
723
XM_017005023.1
XP_016860512.1
7
981
XM_017005024.1
XP_016860513.1
7
723
XM_017005025.1
XP_016860514.1
7
723
XM_017005026.1
XP_016860515.1
7
922
GenBank mRNA
AK022646.1
-
5
507
AK309395.1
-
712
AY219922.1
-
7
684
AY929062.1
-
7
677
BC004514.2
-
3
227
BC065271.1
-
6
637
6165433
BX537956.1
-
7
721
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0