Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288968.1 | NP_001275897.1 | 10 | 1260 | ||
NM_001288969.1 | NP_001275898.1 | 9 | 1146 | |||
NM_001288970.1 | NP_001275899.1 | 8 | 1056 | |||
NM_001288971.1 | NP_001275900.1 | 7 | 860 | |||
NM_001288972.1 | NP_001275901.1 | 9 | 1102 | |||
NM_025149.5 | NP_079425.3 | 9 | 1185 | |||
XM_006722110.3 | XP_006722173.1 | 9 | 1125 | |||
GenBank mRNA | AK024573.1 | - | 9 | 1092 | ||
AK225044.1 | - | 9 | 1092 | |||
AK225561.1 | - | 9 | 1091 | |||
AK294205.1 | - | 10 | 1202 | |||
AK295258.1 | - | 9 | 1054 | |||
AK300145.1 | - | 8 | 964 | |||
AK300625.1 | - | 7 | 860 | |||
AY358660.1 | - | 9 | 1121 | |||
BC012053.1 | - | 9 | 1098 | 4577089 | ||
BC014123.2 | - | 9 | 1105 | 4560029 |