Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025161.5 | NP_079437.5 | 3 | 884 | ||
NR_033338.1 | 3 | 1080 | ||||
XM_006722111.2 | XP_006722174.1 | 3 | 879 | |||
XM_006722112.3 | XP_006722175.1 | 3 | 1030 | |||
XM_017025162.1 | XP_016880651.1 | 3 | 905 | |||
XM_017025163.1 | XP_016880652.1 | 3 | 801 | |||
GenBank mRNA | BC008883.2 | - | 3 | 1053 | ||
BC117139.1 | - | 1 | 404 | 40125690 | ||
BC117141.1 | - | 1 | 404 | 40125692 | ||
BC143861.1 | - | 1 | 404 | |||
DQ989324.1 | - | 3 | 866 |