Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025227.2 | NP_079503.1 | 4 | 457 | ||
GenBank mRNA | AF465765.1 | - | 3 | 240 | ||
AK027068.1 | - | 4 | 434 | |||
AK172819.1 | - | 4 | 436 | |||
AK304064.1 | - | 4 | 362 | |||
AY358723.1 | - | 4 | 365 | |||
BC034415.1 | - | 4 | 311 | 5173032 | ||
BP333980.1 | - | 4 | 456 |