Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271539.1 | NP_001258468.1 | 18 | 1744 | ||
NR_073206.1 | 21 | 2204 | ||||
NR_073207.1 | 19 | 2085 | ||||
XM_006723646.2 | XP_006723709.1 | 20 | 2088 | |||
XM_006723647.2 | XP_006723710.1 | 20 | 2085 | |||
XM_006723648.2 | XP_006723711.1 | 19 | 2031 | |||
XM_006723650.2 | XP_006723713.1 | 19 | 1912 | |||
XM_006723651.2 | XP_006723714.1 | 19 | 1915 | |||
XM_006723652.2 | XP_006723715.1 | 18 | 1749 | |||
XM_006723654.3 | XP_006723717.1 | 11 | 1103 | |||
XM_011529374.1 | XP_011527676.1 | 19 | 2028 | |||
XM_011529375.1 | XP_011527677.1 | 20 | 1977 | |||
XM_011529376.1 | XP_011527678.1 | 19 | 1920 | |||
XM_011529377.1 | XP_011527679.1 | 21 | 2297 | |||
XM_011529378.1 | XP_011527680.1 | 18 | 1806 | |||
XM_011529379.1 | XP_011527681.1 | 18 | 1839 | |||
XM_017028084.1 | XP_016883573.1 | 9 | 1025 | |||
GenBank mRNA | AK302170.1 | - | 18 | 1741 | ||
AL137678.1 | - | 19 | 1969 | |||
DA857535.1 | - | 5 | 387 |