Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024736.1 | NP_001019907.1 | 7 | 1705 | ||
NM_025240.2 | NP_079516.1 | 5 | 1051 | |||
XM_005254700.4 | XP_005254757.1 | 7 | 1823 | |||
XM_011522095.2 | XP_011520397.1 | 7 | 1694 | |||
XM_011522096.2 | XP_011520398.1 | 7 | 1634 | |||
XM_017022638.1 | XP_016878127.1 | 7 | 1623 | |||
GenBank mRNA | AF302102.1 | - | 4 | 817 | ||
AJ583695.1 | - | 6 | 1514 | |||
AK074849.1 | - | 7 | 1595 | |||
AK074997.1 | - | 5 | 1201 | |||
AK075549.1 | - | 7 | 1633 | |||
AK291721.1 | - | 7 | 1542 | |||
AK292711.1 | - | 6 | 1466 | |||
AK296358.1 | - | 7 | 1423 | |||
AK298522.1 | - | 4 | 536 | |||
AL360136.1 | - | 2 | 379 | |||
AY358343.1 | - | 5 | 968 |