Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077497.2 | NP_001070965.1 | 3 | 1984 | ||
NM_025263.3 | NP_079539.2 | 4 | 2047 | |||
GenBank mRNA | AK074531.1 | - | 3 | 1413 | ||
AK131382.1 | - | 2 | 3375 | |||
U63336.1 | - | 4 | 1870 |