Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317237.1 | NP_001304166.1 | 10 | 1100 | ||
NM_001908.4 | NP_001899.1 | 10 | 1131 | |||
NM_147780.3 | NP_680090.1 | 12 | 1293 | |||
NM_147781.3 | NP_680091.1 | 11 | 1250 | |||
NM_147782.3 | NP_680092.1 | 11 | 1219 | |||
NM_147783.3 | NP_680093.1 | 11 | 1205 | |||
XM_006716244.2 | XP_006716307.1 | 12 | 1317 | |||
XM_006716245.2 | XP_006716308.1 | 10 | 1421 | |||
XM_011543812.2 | XP_011542114.1 | 12 | 1226 | |||
XM_017013097.1 | XP_016868586.1 | 11 | 1207 | |||
XM_017013098.1 | XP_016868587.1 | 11 | 1207 | |||
XM_017013099.1 | XP_016868588.1 | 12 | 1249 | |||
XM_017013100.1 | XP_016868589.1 | 11 | 1151 | |||
XM_017013101.1 | XP_016868590.1 | 7 | 674 | |||
GenBank mRNA | AK075393.1 | - | 10 | 1107 | ||
AK092070.1 | - | 11 | 1142 | |||
AK097384.1 | - | 5 | 1061 | |||
AK128472.1 | - | 1 | 1724 | |||
AK130184.1 | - | 10 | 1068 | |||
AK290239.1 | - | 10 | 1068 | |||
AK296845.1 | - | 9 | 872 | |||
AK297687.1 | - | 8 | 832 | |||
BC010240.1 | - | 11 | 1148 | 4154307 | ||
BC095408.1 | - | 10 | 1068 | 30334082 | ||
BX647765.1 | - | 10 | 991 | |||
BX647952.1 | - | 10 | 1071 | |||
BX648239.1 | - | 5 | 6842 | |||
L16510.1 | - | 11 | 1133 | |||
M13230.1 | - | 6 | 566 |