Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201575.1 | NP_001188504.1 | 5 | 619 | ||
NM_001333.3 | NP_001324.2 | 5 | 822 | |||
GenBank mRNA | AB001928.1 | - | 5 | 645 | ||
AK310645.1 | - | 5 | 1091 | |||
AK310740.1 | - | 5 | 754 | |||
AK312806.1 | - | 5 | 619 | |||
AY358641.1 | - | 5 | 822 | |||
BC023504.2 | - | 5 | 636 | 3139314 | ||
BC110512.1 | - | 5 | 691 | 40031744 | ||
Y14734.1 | - | 5 | 641 |