Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCSTAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257317.1 | NP_001244246.1 | 2 | 822 | ||
NM_030788.3 | NP_110415.1 | 2 | 822 | |||
XM_011517321.1 | XP_011515623.1 | 3 | 1191 | |||
XM_011517324.1 | XP_011515626.1 | 3 | 1191 | |||
XM_017013878.1 | XP_016869367.1 | 4 | 1290 | |||
XM_017013879.1 | XP_016869368.1 | 4 | 1289 | |||
XM_017013880.1 | XP_016869369.1 | 4 | 1289 | |||
GenBank mRNA | AF277290.1 | - | 2 | 800 | ||
AF305068.1 | - | 2 | 794 | |||
AK095417.1 | - | 2 | 792 | |||
BC069349.1 | - | 2 | 792 | 7262322 | ||
BC112018.1 | - | 1 | 766 | 8327486 | ||
BC112020.1 | - | 1 | 766 | 8327488 | ||
BC171732.1 | - | 1 | 766 | 9052319 |