Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 17 | 2341 | ||
NM_030793.4 | NP_110420.3 | 18 | 2851 | |||
XM_005268513.1 | XP_005268570.1 | 18 | 3065 | |||
XM_006714797.1 | XP_006714860.1 | 18 | 2939 | |||
XM_011537684.2 | XP_011535986.1 | 18 | 3118 | |||
XM_017009899.1 | XP_016865388.1 | 19 | 3213 | |||
XM_017009900.1 | XP_016865389.1 | 19 | 3277 | |||
XM_017009901.1 | XP_016865390.1 | 18 | 2905 | |||
XM_017009902.1 | XP_016865391.1 | 18 | 2893 | |||
GenBank mRNA | AF177339.1 | - | 4 | 997 | ||
AF251055.1 | - | 11 | 1789 | |||
AK308414.1 | - | 17 | 2290 | |||
AK313558.1 | - | 5 | 1155 | |||
AL832841.1 | - | 8 | 1543 | |||
BC005849.2 | - | 8 | 1650 | |||
BC005873.2 | - | 8 | 1650 | |||
BC015366.1 | - | 4 | 722 | |||
BC033454.1 | - | 18 | 2784 | 4510274 | ||
BC050424.1 | - | 16 | 2246 | 6472693 | ||
BC056147.1 | - | 4 | 902 | |||
BC078159.1 | - | 4 | 742 |