Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271723.1 | NP_001258652.1 | 21 | 2927 | ||
NM_030793.4 | NP_110420.3 | 22 | 3437 | |||
XM_005268513.1 | XP_005268570.1 | 22 | 3651 | |||
XM_006714797.1 | XP_006714860.1 | 22 | 3525 | |||
XM_011537684.2 | XP_011535986.1 | 22 | 3704 | |||
XM_017009899.1 | XP_016865388.1 | 23 | 3799 | |||
XM_017009900.1 | XP_016865389.1 | 23 | 3863 | |||
XM_017009901.1 | XP_016865390.1 | 22 | 3491 | |||
XM_017009902.1 | XP_016865391.1 | 22 | 3479 | |||
GenBank mRNA | AF177339.1 | - | 8 | 1583 | ||
AF251055.1 | - | 15 | 2375 | |||
AK308414.1 | - | 21 | 2876 | |||
AK313558.1 | - | 9 | 1741 | |||
AL832841.1 | - | 12 | 2129 | |||
BC005849.2 | - | 12 | 2236 | |||
BC005873.2 | - | 12 | 2236 | |||
BC015366.1 | - | 8 | 1308 | |||
BC033454.1 | - | 22 | 3370 | 4510274 | ||
BC050424.1 | - | 20 | 2832 | 6472693 | ||
BC056147.1 | - | 8 | 1488 | |||
BC078159.1 | - | 8 | 1328 |