Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281441.1 | NP_001268370.1 | 2 | 524 | ||
NM_030798.4 | NP_110425.2 | 2 | 524 | |||
NM_148842.2 | NP_683682.1 | 2 | 524 | |||
GenBank mRNA | AF410455.1 | - | 2 | 380 | ||
AK309117.1 | - | 2 | 433 | |||
AL136804.1 | - | 2 | 431 | |||
AL536085.3 | - | 2 | 522 | |||
BC007823.2 | - | 2 | 436 | 4304040 | ||
BC019008.2 | - | 2 | 436 | 4330507 | ||
BC032712.1 | - | 2 | 455 | 5495794 | ||
BC040695.1 | - | 2 | 506 | 5785681 |