Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145214.2 | NP_660215.1 | 6 | 1537 | ||
XM_011544285.2 | XP_011542587.1 | 5 | 1373 | |||
XM_017002412.1 | XP_016857901.1 | 6 | 1601 | |||
GenBank mRNA | AF220125.1 | - | 1 | 102 | ||
AF327056.1 | - | 6 | 1537 | |||
AK097825.1 | - | 6 | 1066 | |||
AK226119.1 | - | 5 | 4617 | |||
AK301859.1 | - | 6 | 993 | |||
AK314539.1 | - | 6 | 1520 | |||
AL360165.1 | - | 1 | 69 | |||
BC011629.2 | - | 4 | 629 | |||
BC069227.1 | - | 6 | 1528 | 4875080 |