Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030918.5 | NP_112180.4 | 5 | 874 | ||
XM_005245509.2 | XP_005245566.1 | 5 | 996 | |||
XM_005245510.3 | XP_005245567.1 | 5 | 606 | |||
XM_005245511.4 | XP_005245568.1 | 5 | 902 | |||
XM_011510024.2 | XP_011508326.1 | 4 | 717 | |||
XM_011510025.2 | XP_011508327.1 | 4 | 575 | |||
XM_011510026.2 | XP_011508328.1 | 5 | 970 | |||
XM_017002417.1 | XP_016857906.1 | 4 | 575 | |||
GenBank mRNA | AB007957.2 | - | 5 | 843 | ||
AY044866.1 | - | 4 | 579 | |||
BC012184.1 | - | 5 | 864 | |||
BC051817.1 | - | 2 | 123 | |||
BC071825.1 | - | 4 | 337 | |||
BC100998.1 | - | 5 | 874 | 40006277 | ||
BC100999.1 | - | 5 | 872 | 40006279 | ||
BC101000.1 | - | 5 | 874 | 40006281 | ||
BC101822.1 | - | 5 | 909 | 8069328 | ||
BC101824.1 | - | 5 | 909 | 8069330 | ||
BC107862.1 | - | 3 | 330 |