Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030928.3 | NP_112190.2 | 6 | 939 | ||
GenBank mRNA | AB053172.1 | - | 6 | 939 | ||
AF070552.1 | - | 4 | 479 | |||
AF321125.1 | - | 6 | 910 | |||
BC000137.2 | - | 6 | 901 | 3048353 | ||
BC008676.1 | - | 6 | 777 | |||
BC008860.2 | - | 6 | 832 | |||
BC009410.1 | - | 6 | 895 | 3613566 | ||
BC014202.2 | - | 2 | 200 | |||
BC021126.1 | - | 4 | 471 | |||
BC049205.1 | - | 4 | 543 |