Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039577.3 | NP_001034666.1 | 3 | 494 | ||
NM_001320153.1 | NP_001307082.1 | 3 | 494 | |||
NM_001320155.1 | NP_001307084.1 | 3 | 494 | |||
NM_030937.4 | NP_112199.2 | 3 | 494 | |||
NR_135154.1 | 3 | 494 | ||||
XM_011542216.2 | XP_011540518.1 | 3 | 546 | |||
XM_011542219.2 | XP_011540521.1 | 1 | 85 | |||
XM_011542221.2 | XP_011540523.1 | 1 | 61 | |||
XM_017002420.1 | XP_016857909.1 | 3 | 492 | |||
XM_017002421.1 | XP_016857910.1 | 3 | 478 | |||
XM_017002422.1 | XP_016857911.1 | 3 | 491 | |||
XM_017002423.1 | XP_016857912.1 | 3 | 546 | |||
GenBank mRNA | AF251294.1 | - | 1 | 130 | ||
AK056120.1 | - | 3 | 467 | |||
AK057133.1 | - | 1 | 61 | |||
AK074112.1 | - | 3 | 465 | |||
AK225758.1 | - | 3 | 489 | |||
AL834441.1 | - | 3 | 425 | |||
AY037150.1 | - | 3 | 468 | |||
AY116620.1 | - | 3 | 483 | |||
BC016333.1 | - | 3 | 474 | |||
BM153748.1 | - | 3 | 494 | |||
DA360436.1 | - | 3 | 493 |