Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320273.1 | NP_001307202.1 | 4 | 708 | ||
NM_001320274.1 | NP_001307203.1 | 3 | 600 | |||
NM_030965.2 | NP_112227.1 | 5 | 1118 | |||
GenBank mRNA | AJ507292.1 | - | 5 | 862 | ||
AK056241.1 | - | 5 | 1085 | |||
AK095758.1 | - | 6 | 1004 | |||
AK295019.1 | - | 6 | 961 | |||
AK300902.1 | - | 4 | 633 | |||
BC001201.1 | - | 5 | 1025 | 3356535 | ||
DA166913.1 | - | 3 | 559 |