Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021118.2 | NP_066941.1 | 4 | 754 | ||
XM_005262086.3 | XP_005262143.1 | 4 | 751 | |||
GenBank mRNA | BC021669.1 | - | 4 | 738 | ||
BC038787.1 | - | 1 | 416 | |||
BC058019.1 | - | 4 | 738 | |||
BC070355.1 | - | 4 | 731 | |||
BC126461.1 | - | 4 | 718 | 8992177 | ||
Z22780.1 | - | 2 | 539 |