- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316967.1 | NP_001303896.1 | 7 | 1098 | ||
NM_031302.4 | NP_112592.1 | 8 | 1233 | |||
XM_011538793.2 | XP_011537095.1 | 8 | 1468 | |||
XM_017019999.1 | XP_016875488.1 | 9 | 1549 | |||
GenBank mRNA | AJ271711.1 | - | 5 | 447 | ||
AK027310.1 | - | 6 | 547 | |||
AK056056.1 | - | 7 | 1104 | |||
AY358763.1 | - | 7 | 637 | |||
BC022343.1 | - | 7 | 854 | 4248949 |