Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256316.1 | NP_001243245.1 | 5 | 1457 | ||
NM_031422.5 | NP_113610.2 | 6 | 1495 | |||
XM_005258362.3 | XP_005258419.1 | 2 | 1043 | |||
XM_017026033.1 | XP_016881522.1 | 5 | 1394 | |||
XM_017026034.1 | XP_016881523.1 | 7 | 1618 | |||
GenBank mRNA | AF239821.1 | - | 6 | 1372 | ||
AF332472.1 | - | 5 | 1286 | |||
AF332473.1 | - | 3 | 1053 | |||
AK093349.1 | - | 5 | 1394 | |||
AY358488.1 | - | 6 | 1474 | |||
BC025764.1 | - | 6 | 1439 | 5205829 | ||
KU178792.1 | - | 5 | 1181 | |||
KU178793.1 | - | 4 | 1143 |