Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM107B SIRNA Assays ›
Gene Symbol:
FAM107B
Gene Name:
family with sequence similarity 107 member B
Gene Aliases:
C10orf45
Chromosome Location:
Chr.10: 14518557 - 14774897 on Build GRCh38
UniGene ID:
Hs.446315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38065
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM107B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282695.1
NP_001269624.1
4
801
NM_001282696.1
NP_001269625.1
3
277
NM_001282697.1
NP_001269626.1
2
211
NM_001282698.1
NP_001269627.1
3
525
NM_001282699.1
NP_001269628.1
2
459
NM_001282700.1
NP_001269629.1
2
177
NM_001282701.1
NP_001269630.1
2
298
NM_001282702.1
NP_001269631.1
3
364
NM_001282703.1
NP_001269632.1
2
231
NM_001320735.1
NP_001307664.1
3
243
NM_001320736.1
NP_001307665.1
3
503
NM_001320737.1
NP_001307666.1
3
297
NM_001320738.1
NP_001307667.1
3
263
NM_001320739.1
NP_001307668.1
2
263
NM_001320740.1
NP_001307669.1
2
55
NM_001320741.1
NP_001307670.1
2
156
NM_031453.3
NP_113641.2
3
735
XM_005252616.3
XP_005252673.1
3
132
XM_011519735.1
XP_011518037.1
4
249
XM_017016746.1
XP_016872235.1
3
123
XM_017016747.1
XP_016872236.1
2
100
XM_017016748.1
XP_016872237.1
2
135
XM_017016749.1
XP_016872238.1
2
197
GenBank mRNA
AK127413.1
-
3
735
AK289959.1
-
3
525
AL136885.1
-
2
221
BC064407.1
-
2
181
BC072452.1
-
3
277
BC144707.1
-
4
609
BF979432.1
-
2
271
DA116139.1
-
2
55
DA269906.1
-
2
81
DA305421.1
-
3
174
DA340804.1
-
3
224
DA390740.1
-
2
218
DA409460.1
-
2
237
DA412617.1
-
3
271
DA543225.1
-
2
60
DA813465.1
-
2
84
DA922675.1
-
2
156
DA954468.1
-
2
177
DA962195.1
-
2
85
DB194372.1
-
2
197
DB198416.1
-
3
209
DB240036.1
-
3
254
DC327230.1
-
3
432
DC332216.1
-
3
525
Back To Top
More Information
Gene Ontology Categories:
Function(s)
sensory perception of sound
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0