Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 27, 28 | 3680 | ||
XM_005254708.4 | XP_005254765.1 | 30, 31 | 4048 | |||
XM_005254710.4 | XP_005254767.1 | 31, 32 | 4186 | |||
XM_005254711.4 | XP_005254768.1 | 30, 31 | 4408 | |||
XM_005254712.4 | XP_005254769.1 | 30, 31 | 4065 | |||
XM_005254713.4 | XP_005254770.1 | 30, 31 | 4962 | |||
XM_005254714.3 | XP_005254771.1 | 29, 30 | 3951 | |||
XM_005254715.2 | XP_005254772.1 | 29, 30 | 4657 | |||
XM_006720717.3 | XP_006720780.1 | 30, 31 | 4782 | |||
XM_017022665.1 | XP_016878154.1 | 30, 31 | 5133 | |||
XM_017022666.1 | XP_016878155.1 | 30, 31 | 3992 | |||
XM_017022667.1 | XP_016878156.1 | 28, 29 | 3854 | |||
XM_017022668.1 | XP_016878157.1 | 30, 31 | 4781 | |||
XM_017022669.1 | XP_016878158.1 | 29, 30 | 4647 | |||
GenBank mRNA | AB002318.2 | - | 13, 14 | 1854 | ||
AB621808.1 | - | 8, 9 | 963 | |||
AF402000.1 | - | 27, 28 | 3680 |