Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031475.2 | NP_113663.2 | 7 | 1408 | ||
XM_005263501.2 | XP_005263558.1 | 7 | 1420 | |||
XM_011542231.1 | XP_011540533.1 | 7 | 1420 | |||
XM_011542232.1 | XP_011540534.1 | 7 | 1420 | |||
XM_011542233.2 | XP_011540535.1 | 6 | 847 | |||
XM_011542237.1 | XP_011540539.1 | 7 | 1420 | |||
XM_011542238.2 | XP_011540540.1 | 7 | 1420 | |||
XM_017002433.1 | XP_016857922.1 | 7 | 1420 | |||
XM_017002434.1 | XP_016857923.1 | 7 | 1420 | |||
GenBank mRNA | AL136880.1 | - | 7 | 1408 | ||
AL834172.1 | - | 4 | 514 | |||
BM713713.1 | - | 5 | 595 |