Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031485.3 | NP_113673.3 | 7 | 1703 | ||
GenBank mRNA | AF337808.1 | - | 7 | 1482 | ||
AK074676.1 | - | 7 | 1671 | |||
AK131055.1 | - | 6 | 909 | |||
AK300225.1 | - | 7 | 1156 | |||
AL713729.1 | - | 7 | 1478 | |||
BC002440.2 | - | 7 | 1478 | 3347366 | ||
BC036281.1 | - | 7 | 1454 | |||
EF011614.1 | - | 7 | 1476 |