Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037290.2 | NP_001032367.2 | 8, 9 | 1197 | ||
NM_001256397.1 | NP_001243326.1 | 8, 9 | 1197 | |||
NM_001256398.1 | NP_001243327.1 | 6, 7 | 1090 | |||
NM_001256400.1 | NP_001243329.1 | 7, 8 | 992 | |||
NM_031938.5 | NP_114144.4 | 8, 9 | 1309 | |||
GenBank mRNA | AB096252.1 | - | 8, 9 | 1163 | ||
AF276432.1 | - | 8, 9 | 1806 | |||
AJ290393.1 | - | 8, 9 | 1159 | |||
AK027801.1 | - | 8, 9 | 1192 | |||
AK091783.1 | - | 6, 7 | 1225 | |||
AK297853.1 | - | 7, 8 | 983 | |||
AK315300.1 | - | 8, 9 | 1301 | |||
BC041656.1 | - | 6, 7 | 1053 | 5175925 |