Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KRTAP17-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031964.1 | NP_114170.1 | 1 | 573 | ||
GenBank mRNA | AJ406952.1 | - | 1 | 573 | ||
BC069078.1 | - | 1 | 571 |